Order Entry
Ireland
ContactUsLinkComponent
 
Silicone Rubber Seal
Catalog # METB51191170
Supplier:  Mettler - Toledo
New Product
Silicone Rubber Seal
Catalog # METB51191170
Supplier:  Mettler - Toledo
New Product

Specifications

  • Ø:
    12 mm
  • For:
    For screw cap GL14
  • Description:
    Seal (septa), 2 mm thickness
  • Material:
    Silicone rubber
  • Pk:
    1

Specifications

About this item

Silicone rubber seal (Septa) for screw cap GL14.