Order Entry
Ireland
ContactUsLinkComponent
EasySep™ Human ILC2 Isolation Kit
Catalog # STMC17782
undefined
EasySep™ Human ILC2 Isolation Kit
Catalog # STMC17782
Specifications
  • Description:
    EasySep™ human ILC2 isolation kit, 2×10⁹ cells
  • Pk:
    1 KIT
Specifications
About this item
Immunomagnetic cell isolation kit using particle release technology.
  • Easy-to-use and column-free
  • Up to 95% purity
  • Facilitates rapid isolation of ILC2s without flow sorting
  • No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Human ILC2 Isolation Kit is designed to isolate Group 2 Innate Lymphoid Cells (ILC2s) from leukapheresis products by immunomagnetic positive selection without the use of a flow sorter.
First, CRTH2+ cells are isolated by column-free immunomagnetic positive selection using antibody complexes and EasySep™ Releasable RapidSpheres™. Then, bound magnetic particles are removed from the EasySep™-isolated CRTH2+ cells, and unwanted non-ILC2s are targeted for depletion using antibody complexes and EasySep™ Dextran RapidSpheres™. The final isolated fraction contains highly purified ILC2s that are immediately ready for downstream applications. Following cell isolation with this EasySep™ kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins or other chemically related ligands.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').