EasySep™ Non-Human Primate Custom Positive Selection Kits
Supplier: STEMCELL Technologies
Certificates
About this item
Immunomagnetic positive selection of non-human primate cells.
The EasySep™ Non-Human Primate Custom Positive Selection Kit is designed to isolate any cell type of interest from fresh or previously frozen peripheral blood mononuclear cells by positive selection. STEMCELL Technologies offers a wide selection of antibodies for custom separation and will work with you to isolate a cell type of interest. Please contact our technical support team for more information.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Frequently Bought Together