About this item
Immunomagnetic positive selection kit.
- Fast and easy-to-use
- Up to 98% purity
- No columns required
- Can be combined with SepMate™ for consistent, high-throughput sample processing
The Complete Kit for Human Whole Blood CD34+ Cells is a simple two-step method that is designed to isolate CD34+ cells from whole blood or buffy coat.
First, hematopoietic progenitor cells are pre-enriched using the RosetteSep™ Human Hematopoietic Progenitor Cell Enrichment Cocktail (15186C) with antibodies recognising CD2, CD3, CD14, CD16, CD19, CD24, CD56, CD66b and CD61 surface markers. CD34+ cells are then selected using the EasySep™ Human CD34 Positive Selection Cocktail (18066C), which contains an antibody recognising CD34. RosetteSep™ binds unwanted cells to red blood cells (RBCs), forming immunorosettes, which sediment during density gradient centrifugation. The pre-enriched fraction containing CD34+ cells is harvested from the interface between the plasma and density gradient medium. The pre-enriched CD34+ cells are then labeled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Isolated CD34+ cells are immediately available for downstream applications.