
Lersivirine 98%
Catalog # 472701000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:white to almost white powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:310.35 g/mol
:GHS08
Storage Temperature:Refrigerator
CAS Number:473921-12-9
Formula:C₁₇H₁₈N₄O₂
MDL Number:MFCD16619314