2-Ethynylbenzaldehyde 97%
New Product
:38846-64-9
:
:C₉H₆O
:MFCD04039985
:130.143 g/mol
:230,3°C
:GHS07
:Ambient
- Appearance:light yellow to light brown crystalline powder
- Infrared spectrum:Conforms
- GC:≥96.0 %
- Melting point:62°C to 66°C
Frequently Bought Together