Order Entry
Ireland
ContactUsLinkComponent
 
2,4,6-Trimethyl-m-phenylenediamine 98%
New Product
 
2,4,6-Trimethyl-m-phenylenediamine 98%
New Product

 

  • Appearance:
    white to yellow to light brown powder or crystals an/or chunks
  • Infrared spectrum:
    Conforms
  • GC:
    ≥97.5 %
  • Melting point:
    89°C to 93°C
  • Titration with HClO4:
    97.5 to 102.5 %

 

Frequently Bought Together