Glecaprevir 98%
Catalog # 469341000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:white to almost white crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
Synonyms: ABT-493, A 1282576, (33R,35S,91R,92R,5S,E)-5-(tert-Butyl)-N-((1R,2R)-2-(difluoromethyl)-1-(((1-methylcyclopropyl)sulphonyl)carbamoyl)cyclopropyl)-14,14-difluoro-4,7-dioxo-2,8,10-trioxa-6-aza-1(2,3)-quinoxalina-3(3,1)-pyrrolidina-9(1,2)-cyclopentanacyclotetradecaphan-12-ene-35-carboxamide
:
:838.87 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1365970-03-1
Formula:C₃₈H₄₆F₄N₆O₉S
MDL Number:MFCD30533436