Amentoflavone 98%
Catalog # 469071000.
Supplier: Thermo Fisher Scientific
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:white to yellow to beige powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
Specification Test Results
About this item
:
:538.464 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1617-53-4
Formula:C₃₀H₁₈O₁₀
MDL Number:MFCD00017470