


Salicylic acid 99+%, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69-72-7
:
Formula:(HO)C₆H₄CO₂H
MDL Number:MFCD00002439
:138.12 g/mol
Boiling Pt:211°C (20mmHg)
Flash Pt:157°C
EINECS:200-712-3
:GHS05
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:white to almost white or colorless powder or crystals
- Infrared spectrum:Conforms
- Titration with NaOH:99.0 to 100.5 %
- Melting point:158°C to 161°C
- Related substances:≤0.1 % (4-Hydroxybenzoic acid)
Specification Test Results
Frequently Bought Together