Order Entry
Ireland
ContactUsLinkComponent
Hydrochloric acid 0,1 mol/L
Hydrochloric acid 0,1 mol/L
Catalog # ACSDCH8006013.1L
Supplier:  CPACHEM
New Product
CAS Number:  
Hydrochloric acid 0,1 mol/L
Catalog # ACSDCH8006013.1L
Supplier:  CPACHEM
New Product
CAS Number:  

Specifications

  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Boiling Pt:100 °C
Density:~1,00055 g/cm³
 :36.46 g/mol
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324