Order Entry
Ireland
ContactUsLinkComponent
Charcoal activated (from coconut)
Charcoal activated (from coconut)
Catalog # SPCMC1221-500GM
Supplier:  Spectrum Chemical
New Product
CAS Number:  
Charcoal activated (from coconut)
Catalog # SPCMC1221-500GM
Supplier:  Spectrum Chemical
New Product
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • MESH SIZE:
    8 - 30

Specification Test Results

About this item

 :
 :12.01 g/mol
CAS Number:7440-44-0
 :64365-11-3
Formula:C
MDL Number:MFCD00133992