Order Entry
Ireland
ContactUsLinkComponent
AlphaTec® 88-394 Natural Rubber Latex Gloves
 :  Ansell
New Product
AlphaTec® 88-394 Natural Rubber Latex Gloves
 :  Ansell
New Product
  • Catalog No:
  • ANSE88-394/7
  • ANSE88-394/8
  • ANSE88-394/9
  • ANSE88-394/10
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Latex / latex-free:
  • Latex
  • Latex
  • Latex
  • Latex
  • Material:
  • Natural rubber latex
  • Natural rubber latex
  • Natural rubber latex
  • Natural rubber latex
  • Finish:
  • Fishscale
  • Fishscale
  • Fishscale
  • Fishscale
  • Cuff style:
  • Pinked
  • Pinked
  • Pinked
  • Pinked
  • Palm thickness:
  • 0,52 mm
  • 0,52 mm
  • 0,52 mm
  • 0,52 mm
  • FDA regulated:
  • +
  • +
  • +
  • +
  • Food contact approved:
  • +
  • +
  • +
  • +
Frequently Bought Together