

Iron(III) chloride 105 g/l in aqueous solution Reag. Ph. Eur. 1037801
Catalog # 87833.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:162.21 g/mol
EINECS:231-729-4
UN:2582
ADR:8,III
:GHS05
:GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005