Order Entry
Ireland
ContactUsLinkComponent
Florisil® for pesticide residue analysis, Supelco®
Florisil® for pesticide residue analysis, Supelco®
Catalog # 46382-500G
Supplier:  Merck
CAS Number:  
Florisil® for pesticide residue analysis, Supelco®
Catalog # 46382-500G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :100.38 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249