Order Entry
Ireland
ContactUsLinkComponent
Magnesium silicate for pesticide residue analysis, Supelco®
Magnesium silicate for pesticide residue analysis, Supelco®
Catalog # 20280-U
Supplier:  Merck
CAS Number:  
undefined
Magnesium silicate for pesticide residue analysis, Supelco®
Catalog # 20280-U
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :100.38 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:215-681-1
CAS Number:1343-88-0
Formula:MgO₃Si
MDL Number:MFCD00078249