Order Entry
Ireland
ContactUsLinkComponent
Organic reference standard, Ethanol absolute 2,000 µg/ml in methanol, Supelco®
Organic reference standard, Ethanol absolute 2,000 µg/ml in methanol, Supelco®
  48075.
 :  Merck
 :  
undefined
Organic reference standard, Ethanol absolute 2,000 µg/ml in methanol, Supelco®
  48075.
 :  Merck
 :  

 

  • Pk:
    1
  • CAS No.:
    64-17-5
  • Description:
    Ethanol absolute 2,000 µg/ml in methanol
  • Formula:
    C₂H₆O
  • Description:
    Ethanol absolute