3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Supplier: Roth Carl
Certificates
About this item
CAS Number:75621-03-3
:
Formula:C₃₂H₅₈N₂O₇S
MDL Number:MFCD00081083
:614.89 g/mol
Merck Index:13,02052
Melting Pt:116 °C
Storage Temperature:Refrigerator
Frequently Bought Together