N,N-Dimethyl-1,3-phenylenediammonium dichloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N1,N1-Dimethylbenzene-1,3-diamine dihydrochloride, N,N-Dimethyl-1,3-phenylenediamine dihydrochloride, N,N-Dimethyl-m-phenylenediamine dihydrochloride
CAS Number:3575-32-4
:
Formula:C₈H₁₄Cl₂N₂
MDL Number:MFCD00012971
Molecular Weight:209.12 g/mol
Melting Pt:216 °C (dec.)
Storage Temperature:Ambient
EINECS:222-693-0
UN:2811
ADR:6.1 III
:GHS06
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together