Genomic DNA Purification Kit
Catalog # STMC79020
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Method / Format:
- Refine target molecule:
- Sample type:
- Target molecule:
- Description:Genomic DNA purification kit
- Environmentally Preferable:
- No. of reactions:250 preparations
Specifications
About this item
For purification of genomic DNA from cells or tissue.
The Genomic DNA Purification Kit uses a membrane-based system to quickly purify high-quality genomic DNA that is suitable for downstream applications such as agarose gel electrophoresis, restriction enzyme digestion, and PCR analysis. Genomic DNA can be purified from cells in as little as 20 minutes. Using a microcentrifuge, up to 20 mg of tissue (mouse tail or animal tissue) or between 1×10⁴ to 5×10⁶ cells can be processed per purification. NOTE: Genomic DNA purification from tissue requires overnight digestion with Proteinase K Solution (Catalog #STMC79016) prior to purification.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').