Showa 257X Cut Resistant Gloves, Uncoated
Supplier: Showa
Certificates
About this item
This cut level A7 aramid glove is designed specifically for use in automotive, stamping, and industrial applications.
- Cut-resistant and abrasion-resistant
- Increased dexterity
- Lightweight and breathable
- Machine washable
- Form-fitting
- Touch screen compatible
A 13 gauge lightweight design offers high dexterity, very high cut resistance, and can be laundered multiple times without compromising the glove’s performance. Thanks to the lightweight and thin construction, the 257X makes a great liner to additional workwear or PPE.
Certifications: Cat II. EN 388:2016+A1:2018, 4X42F. EN 407:2020, X1XXXX.
Packaging: Gloves are packaged 12 pair per polybag, 6 polybags per case (72 total pair per case).
Specifications
- Catalog No:
- BEST257X-8/L
- BEST257X-7/M
- BEST257X-6/S
- BEST257X-10/XXL
- BEST257X-9/XL
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Material:
- Aramid
- Aramid
- Aramid
- Aramid
- Aramid
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile