Order Entry
Ireland
ContactUsLinkComponent
Propidium iodide cell viability stain
Propidium iodide cell viability stain
Catalog # STMC75002
CAS Number:  
undefined
Propidium iodide cell viability stain
Catalog # STMC75002
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :668.4 g/mol
CAS Number:25535-16-4
Formula:C₂₇H₃₄I₂N₄
MDL Number:MFCD00011921