Order Entry
Ireland
ContactUsLinkComponent
GemCell™ Plus Xeno-Free Human Serum, World Grade, Gamma Irradiated
GemCell™ Plus Xeno-Free Human Serum, World Grade, Gamma Irradiated
  GEMB100-916G-100
New Product
undefined
GemCell™ Plus Xeno-Free Human Serum, World Grade, Gamma Irradiated
  GEMB100-916G-100
New Product

 

  • Serum format:
    Frozen
  • Sterility:
    Sterile filtered
  • Description:
    GemCell™ Plus xeno-free, world grade human serum AB - gamma irradiated
  • Shipping conditions:
    Ships on dry ice
  • Origin:
    US
  • Storage temperature:
    Store frozen at −20...−10 °C
  • Pk Info:
    200×100 ml
  • Pack type:
    Bottle
  • Pk:
    100 mL

 

 

Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.

  • This product was developed without the use of animal material
  • Custom fill sizes in bottle and bag format offered
  • 0,1 µm sterile-filtered
  • Supports clinical and commerical applications
  • Meet US and EU regulatory requirements
  • Gamma irradiated