GemCell™ Plus Xeno-Free Human Serum, World Grade
GEMB100-916-100
New Product
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:GemCell™ Plus xeno-free, world grade human serum AB
- Shipping conditions:Ships on dry ice
- Origin:US
- Storage temperature:Store frozen at −20...−10 °C
- Pk Info:200×100 ml
- Pack type:Bottle
- Pk:100 mL
Product has minimum order quantity of 200 units. GemCell™ Plus Xeno-Free, world grade is collected from a maximum of 16 healthy male donors of the AB serotype. Product is defibrinated from source plasma using therapeutic grade recombinant human thrombin. Based on extensive testing and manufacturing process that is Xeno-Free, supports clinical and commercial applications.
- This product was developed without the use of animal material
- Custom fill sizes in bottle and bag format offered
- 0,1 µm sterile-filtered
- Supports clinical and commerical applications
- Meet US and EU regulatory requirements