Order Entry
Ireland
ContactUsLinkComponent
Speirs Robertson AMS Anti-Vibration Platform
Speirs Robertson AMS Anti-Vibration Platform
  COLE11500-20
New Product
Speirs Robertson AMS Anti-Vibration Platform
  COLE11500-20
New Product
  • Description:
    AMS anti-vibration platform, 750×750 mm, 440 kg
  • Pk:
    1
Improves performance of sensitive instruments by shielding them from vibrations.
  • High performance isolators available in a choice of performance levels
  • Suitable for instruments from 10 to 200 kg
ave time and money by making sure your expensive high performance instruments are not underperforming due to a poor working environment. Vibrations limit the performance of sensitive equipment but our platforms remove these vibrations allowing you to get the performance you expect.
Platforms are simple to install, easy to use and maintenance free.