CRYO-LNG™ Gloves
Supplier: TEMPSHIELD
New Product
Certificates
About this item
Hand protection for industrial settings working with LNG or liquid nitrogen.
- Abrasion resistant palm fabric safeguards against rough surfaces
- A thin, seamless 100% waterproof liner provides protection from spills and splashes
- Cryogenic protection for ultra-cold applications down to –196 °C (–320 °F)
- High performance, thermal inner lining wicks moisture away from hands, maintaining comfort over extended periods
Gloves are designed to provide a high level of thermal and splash protection where the exposure to cryogenic fluids might exist.
State-of-the art materials and a multi-layered construction allow for a maximum level of thermal protection, flexibility, and dexterity which are essential features when function is important and safety is critical.
Applications: Delivering cryogenic gases, servicing cryogenic systems, refueling and servicing at LNG filling stations, working in LNG transportation and facilities, working in mining, oil, and gas.
Certifications: EN 511, EN 388, EN ISO 4120
Caution: Not intended for immersion in liquid nitrogen or other cryogenic liquids.
Specifications
- Catalog No:
- TEMPCLMXWP
- TEMPCLMMWP
- TEMPCLMXXWP
- TEMPCLMLWP
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable