Supelco® PESTANAL® Organic Reference Standard, Spirotetramat
Catalog # 32713-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:PESTANAL®, Analytical standard
- CAS No.:203313-25-1
- Description:Spirotetramat
- Seq:05
- Storage conditions:2...30 °C
- Hazard class description:H317 - H319 - H335 - H361fd - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):cis-3-(2,5-Dimethylphenyl)-8-methoxy-2-oxo-1-azaspiro[4.5]dec-3-en-4-yl ethyl carbonate; cis-4-(Ethoxycarbonyloxy)-8-methoxy-3-(2,5-xylyl)-1-azaspiro[4.5]dec-3-en-2-one
- Formula:C₂₁H₂₇NO₅
- Molecular weight:373,44
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS07
Spirotetramat may be used as a reference standard for QuEChERS based methodology for the determination of spirotetramat and its relevant metabolites in pistachio samples by liquid chromatography-tandem mass spectrometry.
Caution: P201 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P308 + P313