Order Entry
Ireland
ContactUsLinkComponent
Magicard™ Prima 8 Re-Transfer Film
Magicard™ Prima 8 Re-Transfer Film
Catalog # BRDY321980
Supplier:  Brady
New Product
undefined
Magicard™ Prima 8 Re-Transfer Film
Catalog # BRDY321980
Supplier:  Brady
New Product

Specifications

  • Colour:
    Transparent
  • Description:
    Re-transfer film, 1000 single-sided cards
  • Length:
    9,75 m
  • Pk:
    1
  • Width:
    60 mm
  • For:
    Prima 8

Specifications

About this item

The Magicard™ Prima 8 transfer film allows users to transfer colours on the cards.

  • Allows for over-the-edge printing
  • Guarantees to print 1000 single-sided cards

Transfer colours on the cards.

Delivery: 1 re-transfer film.