Order Entry
Ireland
ContactUsLinkComponent
Cell Navigator™ Flow Cytometric Exosome Staining Kit
Cell Navigator™ Flow Cytometric Exosome Staining Kit
Catalog # AATB22426
Supplier:  AAT Bioquest
New Product
CAS Number:  
Cell Navigator™ Flow Cytometric Exosome Staining Kit
Catalog # AATB22426
Supplier:  AAT Bioquest
New Product
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Tests per kit:
    100 tests
Specifications
About this item
Designed for consistent and dependable results. The Cell Navigator™ Flow Cytometric Exosome Staining Kit addresses these challenges by enabling the detection of CD9-positive exosomes in purified or bead-bound samples.
  • Rapid and accurate exosome detection in diverse sample matrices, including cell culture supernatants, biological fluids, and isolated exosome preparations
  • Six-month shelf life
Exosomes, nanoscale (30 to 150 nm) membrane-bound vesicles secreted by cells, are central to intercellular communication.
Delivery: Shipped with blue ice.
Caution: To ensure effective detection and analysis, it is essential to enrich the cell culture media for exosomes before staining. Research use only.