Amplite® Fluorimetric DPP4 Inhibitor Screening Kit
Catalog # AATB11324
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Pk:100 Tests
- Tests per kit:100 tests
Specifications
About this item
Designed for consistent and dependable results. The Amplite® Fluorimetric DPP4 Inhibitor Screening Kit provides an efficient and direct method for evaluating potential DPP4 inhibitors.
- Ex/Em=360/460 nm
- Assay is optimised for high-throughput screening systems
This kit includes Sitagliptin, a well-characterised DPP4 inhibitor used in diabetes treatment, as a reference control, facilitating the assessment of inhibitor efficacy.
Delivery: Shipped with blue ice.
Caution: Research use only.