Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Demeton-S-Methyl-Sulfon
  07313-100MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Demeton-S-Methyl-Sulfon
  07313-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • CAS No.:
    17040-19-6
  • Description:
    Demeton-S-methyl-sulfon
  • Environmentally Preferable:
  • Storage conditions:
    2...8 °C
  • Hazard class description:
    H300 + H330 - H311 - H411
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00055415
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    262,28

 

 

 : This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
 : P260 - P264 - P273 - P280 - P302 + P352 + P312 - P304 + P340 + P310