Order Entry
Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Anilazine
Supelco® TraceCERT® Organic Reference Standard, Anilazine
Catalog # CRM05691-25MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Anilazine
Catalog # CRM05691-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    101-05-3
  • Description:
    Anilazine
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H410
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00041815
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    6-(2-Chloroanilino)-2,4-dichloro-1,3,5-triazine
  • Molecular weight:
    275,52

Specifications

About this item

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find the following uses:

  • Analysis of long-term persistence of three 14C-labeled pesticides- ethidimuron, methabenzthiazuron, anilazine, and their metabolites in soil
  • Gas chromatographic-mass spectrometric (GC-MS) determination of different classes of pesticide residues from various wastewater samples obtained from culturing and processing of potatoes
Caution: P264 - P273 - P280 - P302 + P352 - P305 + P351 + P338 - P332 + P313