Supelco® TraceCERT® Organic Reference Standard, Fenobucarb
Catalog # CRM05583-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:3766-81-2
- Description:Fenobucarb
- Storage conditions:–20 °C
- Hazard class description:H302 - H410
- Shipping conditions:STD
- MDL No.:MFCD00053081
- M-Clarity quality level:MQ300
- Synonym(s):BPMC
- Molecular weight:207,27
Specifications
About this item
Fenobucarb may be used as reference material for the following:
- Method development and validation for simultaneous determination of fenobucarb enantiomers in vegetables crops soils and water by UPLC-MS/MS using a Daicel IG-3 chiral column
- Method development for determination of fenobucarb residues in vegetable samples using screen-printed carbon electrode modified with graphene nanoribbons-ionic liquid-cobalt phthalocyanine composites based on flow injection analysis (FIA)
- Quantification of fenobucarb residues in animal (porcine muscle egg and whole milk) and aquatic (eel flatfish and shrimp) food products using QuEChERS extraction and liquid chromatography-electrospray ionisation tandem mass spectrometry (LC-ESI/MS-MS)
- Development of an electrochemical screening assay for the detection of 4 phenyl carbamates in grains samples and comparing the data with UHPLC-MS/MS
- Quantitative analysis of fenobucarb residues in beef muscles using QuEChERS method and liquid chromatography-tandem mass spectrometry
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501