Order Entry
Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Diclofop-methyl
Supelco® TraceCERT® Organic Reference Standard, Diclofop-methyl
Catalog # CRM04291-100MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Diclofop-methyl
Catalog # CRM04291-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    51338-27-3
  • Description:
    Diclofop-methyl
  • Storage conditions:
    15...25 °C
  • Hazard class description:
    H302 - H317 - H410
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00128052
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    341,19

Specifications

About this item

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The diclofop-methyl CRM may find its use as described below:

  • To study the enantioselective degradation of diclofop-methyl and diclofop in two soil samples under aerobic and anaerobic conditions using high-performance liquid chromatography (HPLC)
  • Simultaneous extraction and determination of enantiomers of diclofop-methyl and diclofop in cole using high-performance liquid chromatography-chiral stationary phase (HPLC-CS)
  • Development of a multi-residue method for simultaneous estimation of 16 pesticides in water samples using direct immersion SPME followed by GC-MS analysis
  • Simultaneous extraction and determination of aryloxyphenoxy-propionate herbicides from water samples by dispersive magnetic solid-phase extraction (d-MSPE) with high-performance liquid chromatography-diode array detector (HPLC-DAD) and ultra-high pressure liquid chromatography -triple quadrupole mass spectrometer (UHPLC-MS/MS)
  • Evaluating the capacity of silica-bonded deoxycholic acid as a stationary phase having a 3,5-dinitrophenylcarbamoyl and a calix[4]arene substituent for enantiomeric separations
Caution: P261 - P264 - P273 - P280 - P301 + P312 - P302 + P352