Order Entry
Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Dichlofluanid
Supelco® TraceCERT® Organic Reference Standard, Dichlofluanid
  CRM04195-100MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Dichlofluanid
  CRM04195-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    1085-98-9
  • Description:
    Dichlofluanid
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H317 - H319 - H332 - H400
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00078639
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    333,23

 

 

It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Dichlofluanid CRM may also find its use as described below:

  • To determine the concentration of dichlofluanid and its metabolite in seawater and sediment of three marinas in Greece before and during the yachting season
  • To evaluate the biochemical and physiological effects of dichlofluanid exposure on the brown mussel Perna perna
  • Photocatalytic degradation of chlorothalonil and dichlofluanid over TiO₂ suspensions
  • Testing toxicity of the fungicides penconazol, cymoxanil, and dichlofluanid and their effects on the specific growth rate, biomass production, respiration rate, and yeast viability
  • To estimate the rates of disappearance of dichlofluanid and tebuconazole following successive applications to two varieties of lettuce grown in greenhouse conditions
 : P261 - P273 - P280 - P302 + P352 - P304 + P340 + P312 - P305 + P351 + P338