Supelco® TraceCERT® Organic Reference Standard, Chlormequat chloride
Catalog # 93757-50MG
Supplier: Merck
CAS Number:
Specifications
- Pk:50 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:999-81-5
- Description:Chlormequat chloride
- Environmentally Preferable:
- Storage conditions:15...25 °C
- Hazard class description:H302 - H311
- MDL No.:MFCD00011869
- M-Clarity quality level:MQ300
- Synonym(s):Chlorocholine chloride; (2-Chloroethyl)trimethylammonium chloride; Chlormequat chloride; Choline dichloride
- Formula:(CH₃)₃N(Cl)CH₂CH₂Cl
- Molecular weight:158,07
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P264 - P270 - P280 - P301 + P312 - P302 + P352 + P312 - P361 + P364