Order Entry
Ireland
ContactUsLinkComponent
 
Supelco® TraceCERT® Organic Reference Standard, Isocarbamide
  91676-50MG
 :  Merck
 :  
Supelco® TraceCERT® Organic Reference Standard, Isocarbamide
  91676-50MG
 :  Merck
 :  

 

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    30979-48-7
  • Description:
    Isocarbamide
  • Storage conditions:
    15...25 °C
  • Shipping conditions:
    STD
  • MDL No.:
    MFCD00055504
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    185,22

 

 

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Isocarbamide CRM may also be used as given below:

  • Analysis of 250 pesticide residues in 6 kinds of unprocessed food products from farms in Poland using modified QuEChERS combined with high-performance liquid chromatography and tandem mass spectrometry (HPLC-MS/MS)
  • Rapid determination of 166 pesticide residues in fruit and vegetable samples at low parts per billion (ppb) concentrations using ultrahigh-performance liquid chromatography and electrospray ionisation quadrupole Orbitrap high-resolution mass spectrometry (UHPLC/ESI Q-Orbitrap) and QuEChERS for sample extraction
  • Development and validation of a method for the screening of 138 pesticide residues in fruit- and vegetable- based infant foods using ultra-performance liquid chromatography-electrospray ionisation quadrupole time-of-flight mass spectrometry (UPLC QqTOF)
  • Multi-residue estimation of 250 pesticide residues in samples of different parts of apples oranges grapefruits and lemons using a modified procedure based on the QuEChERS method and liquid chromatography-tandem mass spectrometry (LC-MS/MS) analysis
  • Development of an LC/electrospray ionisation (ESI)-MS/MS and ultra-HPLC (UHPLC)/quadrupole (Qq)-time-of-flight (TOF)-MS based procedure for the determination of 141 pesticide residues from 6 types of loose tea samples
  • Simultaneous estimation of 148 pesticide residues in berry fruit samples using liquid chromatography-electrospray ionisation tandem mass spectrometry (LC/ESIMS/MS) and ultrahigh-pressure liquid chromatography-electrospray ionisation quadrupole timeof-flight mass spectrometry (UHPLC QqTOF MS)