Supelco® TraceCERT® Organic Reference Standard, Isazophos
Catalog # 91607-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:42509-80-8
- Description:Isazophos
- Storage conditions:–20 °C
- Hazard class description:H300 + H310 + H330 - H317 - H373 - H410
- Shipping conditions:WICE
- MDL No.:MFCD00078699
- M-Clarity quality level:MQ300
- Molecular weight:313,74
Specifications
About this item
It is intended to be used as a certified reference material (CRM) for calibration in chromatography and other analytical techniques. Isazophos CRM may also find its use as described below:
- To develop a GC/MS procedure utilising selected ion monitoring (SIM) for quantification of isazophos in bovine liver kidney muscle and fat at various times after exposure
- To develop multiresidue method for quantification of 19 metabolites of organophosphorus pesticides synthetic pyrethroids selected herbicides and DEET (NN-diethyl-m-toluamide) in human urine using LC-MS
- To determine the efficacy of isazophos against Japanese beetle (Popillia japonica Newman) grubs using two subsurface placement technology high-pressure liquid injection (HPLI) and an adapted overseeder (OS) applied to golf course roughs
Caution: P262 - P273 - P280 - P302 + P352 + P310 - P304 + P340 + P310 - P314