Order Entry
Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Iprobenfos
Supelco® TraceCERT® Organic Reference Standard, Iprobenfos
Catalog # 91386-100MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Iprobenfos
Catalog # 91386-100MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    26087-47-8
  • Description:
    Iprobenfos
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H302 - H411
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD00191226
  • M-Clarity quality level:
    MQ300
  • Molecular weight:
    288,34

Specifications

About this item

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The iprobenfos CRM may also find following uses:

  • Sensitive detection of iprobenfos and edifenphos using gold nanoparticle AuNP-based colorimetric multi-aptasensor from rice samples
  • To investigate light-induced catalytic degradation of iprobenfos fungicide using poly(3-octylthiophene-25-diyl) film by varying oxygen pressure of bubbling gases
  • To determine the variation in sensitivity of Magnaporthe oryzae isolates from different geographic areas towards edifenphos and iprobenfos fungicides
  • To study the response of zooplankton community against the application of simetryn and iprobenfos using natural zooplankton community in microcosm tanks
  • To evaluate and compare the virulence of Magnaporthe oryzae isolates sensitive and resistant to edifenphos and iprobenfos on rice plants treated with or without fungicides
Caution: P264 - P270 - P273 - P301 + P312 - P391 - P501