Supelco® TraceCERT® Organic Reference Standard, Heavy metal mix IX
Catalog # 89471-100ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:12% nitric acid
- Pk:100 mL
- Standards:TraceCERT®, certified reference material
- Description:Heavy metal mix IX
- Environmentally Preferable:
- Storage conditions:No temperature limit
- Hazard class description:H290 - H314 - H350 - H412
- Shipping conditions:STD
- M-Clarity quality level:MQ300
Specifications
About this item
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Caution: P202 - P273 - P280 - P303 + P361 + P353 - P304 + P340 + P310 - P305 + P351 + P338