Supelco® TraceCERT® Organic Reference Standard, Thiofanox
Catalog # 77068-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Certified reference material, TraceCERT®
- CAS No.:39196-18-4
- Description:Thiofanox
- Storage conditions:–20 °C
- Hazard class description:H300 + H310 - H410
- Shipping conditions:DICE
- MDL No.:MFCD00055322
- Molecular weight:218,32
Specifications
About this item
The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Thiofanox CRM may also be used as given below:
- 126 pesticide residues in chicken samples using modified QuEChERS for sample pre-treatment and ultra-high-performance liquid chromatography combined with quadrupole time-of-flight mass spectrometry (UHPLC-QToF-MS) for quantification
- Multi-residue analysis of 423 pesticides consisting of their isomers and metabolites in 124 green tea C. sinensis nutraceutical products by two-dimensional gas chromatography coupled with time-of-flight mass spectrometry
- QuEChERS based extraction and ultra-high performance liquid chromatography-triple quadrupole tandem mass spectrometric (UHPLC-QqQ-MS/MS) analysis of LC-amenable pesticides in green tea (C. sinensis) based nutraceutical products
- Multi-residue analysis of 80 pesticide residues in 30 aquaculture water samples by single step solid phase extraction (SPE)-cleanup and ultra-high performance liquid chromatography-Q/Orbitrap mass spectrometry
Caution: P262 - P264 - P273 - P280 - P301 + P310 - P302 + P352 + P310