Order Entry
Ireland
ContactUsLinkComponent
Supelco® TraceCERT® Organic Reference Standard, Tepraloxydim
Supelco® TraceCERT® Organic Reference Standard, Tepraloxydim
Catalog # 75974-50MG
Supplier:  Merck
CAS Number:  
Supelco® TraceCERT® Organic Reference Standard, Tepraloxydim
Catalog # 75974-50MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Certified reference material, TraceCERT®
  • CAS No.:
    149979-41-9
  • Description:
    Tepraloxydim
  • Storage conditions:
    –20 °C
  • Hazard class description:
    H351 - H361fd
  • Shipping conditions:
    WICE
  • MDL No.:
    MFCD03792787
  • M-Clarity quality level:
    MQ300
  • Synonym(s):
    trans-2-[1-(3-Chloroallyloxyimino)propyl]-3-hydroxy-5-(tetrahydro-2H-pyran-4-yl)-2-cyclohexen-1-one
  • Molecular weight:
    341,83

Specifications

About this item

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. Tepraloxydim CRM may also be used as given below:

  • Screening and quantification of 569 pesticides in honeysuckle samples by a combination of ultrahigh-performance liquid chromatography (UHPLC) and quadrupole-Orbitrap high-resolution mass spectrometry
  • Detection of 252 pesticides extracted from surface water samples by solid-phase extraction (SPE) using liquid chromatography quadrupole-orbitrap high-resolution tandem mass spectrometry
  • Evaluation of 250 pesticide residues extracted by QuEChERS method from 103 processed fruit samples using ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS)
  • Development of a method for simultaneous quantification of 200 pesticide residues in lettuce and orange samples based on methanol as the extraction solvent ultra-high-performance liquid chromatography (UHPLC) for separation and tandem mass spectrometry (MS/MS) for detection
  • Residue analysis of 261 pesticides their metabolites and polychlorinated biphenyls using modified QuEChERS protocol; optimised for extremely low molecular weight samples and liquid/gas chromatography coupled with tandem mass spectrometry
  • To develop and validate a method for the quantitative analysis of 207 pesticide residues in honey samples using both LC-MS/MS and GC-MS/MS compounds
Caution: P201 - P202 - P280 - P308 + P313 - P405 - P501