Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
iFluor® 670 styramide
iFluor® 670 styramide [Replacement for Opal 690]
iFluor® 670 styramide [Replacement for Opal 690]
iFluor® 670 styramide [Replacement for Opal 690]
Catalog # AATB45047
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
iFluor® 670 styramide [Replacement for Opal 690]
Catalog # AATB45047
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
100
Seq:
0100
Pk Info:
100 Slides
Specifications
About this item
Product Details
:
Product Details