0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Biotin-PEG4-amine
Biotin-PEG4-amine
Biotin-PEG4-amine
Biotin-PEG4-amine
Catalog # AATB3013
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Biotin-PEG4-amine
Catalog # AATB3013
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
5 mg
Seq:
2210
Specifications
About this item
Product Details
:
:
462.606 g/mol
CAS Number:
663171-32-2
Formula:
C₂₀H₃₈N₄O₆S
MDL Number:
MFCD26142992
Product Details