Anti-Adenylate Kinase 1 Rabbit Polyclonal Antibody
Catalog # ANTIA95852-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Adenylate Kinase 1
- Antigen symbol:Adenylate Kinase 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P00568
- Antigen synonyms:ATP AMP transphosphorylase|Ak1|Myokinase|KAD1_HUMAN|Adenylate kinase isoenzyme 1|KAD1|AK 1|Adenylate kinase soluble|ATP-AMP transphosphorylase 1
- Storage buffer:Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
- Molecular weight:21 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human KAD1 (amino acids 101-150)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to KAD1 for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, IF, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: Adenylate Kinase 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat