Order Entry
Ireland
ContactUsLinkComponent
 
Anti-ATF1 Rabbit Polyclonal Antibody
  ANTIA96418-100
New Product
undefined
Anti-ATF1 Rabbit Polyclonal Antibody
  ANTIA96418-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Activating transcription factor 1
  • Antigen symbol:
    ATF1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P18846
  • Antigen synonyms:
    cAMP dependent transcription factor 1|ATF1_HUMAN|ATF1 FUS fusion gene|ATF1|cAMP-dependent transcription factor ATF-1|ATF1 EWS fusion gene|Cyclic AMP dependent transcription factor ATF1|Cyclic AMP dependent transcription factor ATF 1|ATF 1
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Molecular weight:
    29 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human ATF1 (amino acids 176-225)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to ATF1 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IF, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:5000

Type: Primary
Antigen: ATF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat