Anti-HEXB Mouse Monoclonal Antibody [clone: HEXB/7762]
Catalog # ANTIA315075-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beta hexosaminidase beta chain
- Antigen symbol:HEXB
- Clonality:Monoclonal
- Clone:HEXB/7762
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P07686
- Antigen synonyms:Beta N acetylhexosaminidase|HCC 7|Cervical cancer proto oncogene 7 protein|Beta-N-acetylhexosaminidase subunit beta|Epididymis luminal protein 248|ENC 1AS|Beta-hexosaminidase subunit beta chain A|Cervical cancer proto-oncogene 7 protein|Beta hexosaminidase subunit beta
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:63 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment human HEXB protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [HEXB/7762] antibody to HEXB for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ml
Type: Primary
Antigen: HEXB
Clonality: Monoclonal
Clone: HEXB/7762
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human