Anti-Biotin Rabbit Recombinant Antibody [clone: BTN/8672R]
Catalog # ANTIA314514-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Biotin
- Antigen symbol:Biotin
- Clonality:Recombinant
- Clone:BTN/8672R
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Species independent
- Western blot:Yes
- Form:Liquid
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:244 Da
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Biotinylated sheep immunoglobulin
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [BTN/8672R] antibody to Biotin for flow cytometry, IF, WB and IHC-P with samples derived from Species Independent.
- Validated applications: flow cytometry, IF, WB, IHC-P
- Recommended dilutions: In situ Nucleic Acid Hybridization flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Biotin
Clonality: Recombinant
Clone: BTN/8672R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Species Independent