Anti-YL1 Rabbit Polyclonal Antibody
Catalog # ANTIA97109-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:CFL1
- Antigen symbol:YL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q15906
- Antigen synonyms:Transcription factor-like 1|Protein YL-1|Swc2|HGNC11644|Transcription factor like 1 (YL1)|Vacuolar protein sorting 72 (yeast)|Transcription Factor Like 1|TCFL1|Vacuolar protein sorting 72 homolog (S. cerevisiae)
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:40 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide derived from human VPS72 (amino acids 131-180)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to VPS72 for WB, IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: YL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse