Order Entry
Ireland
ContactUsLinkComponent
Anti-ARPP21 Rabbit Polyclonal Antibody
Anti-ARPP21 Rabbit Polyclonal Antibody
Catalog # ANTIA96960-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-ARPP21 Rabbit Polyclonal Antibody
Catalog # ANTIA96960-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    0710001E13Rik
  • Antigen symbol:
    ARPP21
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9UBL0
  • Antigen synonyms:
    Cyclic AMP-regulated phosphoprotein, 21|ARPP21|cAMP-regulated phosphoprotein, 21-KD|Cyclic AMP-regulated phosphoprotein|ARP21_HUMAN|cAMP-regulated phosphoprotein 21|cAMP regulated phosphoprotein, 21kDa|AI853636|ARPP-21
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    89 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at +4 °C
  • Immunogen:
    Synthetic peptide derived from human ARPP21 (amino acids 211-260)
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to ARPP21 for WB, IHC and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, IHC, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:5000

Type: Primary
Antigen: ARPP21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse